Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide (+99% Purity), MP Biomedicals™ PROMO

Catalog No. ICN9018690P Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
0.1 L
500 mL
1 L

Catalog No. ICN9018690P

Supplier: MP Biomedicals, Inc 0219018690FP

Dimethyl Sulfoxide is used to enhance dermal absorption of many chemicals.

  • Dimethyl sulfoxide is used to enhance dermal absorption of many chemicals.
  • A solvent for many organic and inorganic compounds including fats, carbohydrates, dyes, resins, and polymers.
  • Used in antifreeze or hydraulic fluids and as a cryopreservative for cell cultures.
  • Used in the oxidation of thiols and disulfides to sulfonic acids.
  • Used as a PCR cosolvent to help improve yields, especially in long PCR.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Density 1.104 g/cm3 at 20 °C
Quantity 500 mL
Assay Percent Range 99+%
Synonym demasorb
Formula Weight 78.13
Chemical Name or Material Dimethyl Sulfoxide

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.