Learn More
Dimethyl sulfoxide, ACS, 99.9% min
Description
It is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. Used in antifreeze or hydraulic fluids. Utilized in the oxidation of thiols and disulfides to sulfonic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 18.4°C |
Color | Colorless |
Density | 1.101 g/mL |
Boiling Point | 189°C |
Quantity | 1 L |
Flash Point | 87°C (189°F) |
Physical Form | Clear,colorless Liquid |
Refractive Index | 1.479 |
Assay Percent Range | 99.9% |
Beilstein | 506008 |
Show More |
Safety and Handling
Hazard Category | H227 |
Precautionary Statement | P210-P370+P378q-P501c |
EINECSNumber | 200-664-3 |
RTECSNumber | PV6210000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.