Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylpyrrolidone, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0%, Honeywell

Catalog No. 6003085 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
4 x 4 L
Packaging:
Poly Bottle
This item is not returnable. View return policy

Catalog No. 600-30-85

Supplier: Honeywell Chemicals LP3044P

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

CAS Max % 100.0000%
Density 1.0259 g/cm3
Boiling Point 202°C
Flash Point 88°C
Packaging Poly Bottle
Quantity 4 x 4 L
Beilstein 106420
Solubility Information Miscible in Water at 20°C
Formula Weight 99.15
Percent Purity >99%
Grade ACS Reagent
Assay by GC Analysis
Chemical Name or Material N-Methylpyrrolidone, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0%
Show More Show Less
ShelfLife 547 days from date of manufacture
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.