Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethyl Sulfoxide-d6, 99.9 Atom% D, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
5 g
10 g
25 g
50 g
10 x 0.75 mL
10 x 1 g
Packaging:
Amber Glass Bottle
Ampule
Description
Spectrum™ Chemical Dimethyl Sulfoxide-d6, 99.9 Atom Percent D - Spectrum offers a broad range of high quality deuterated products for all your routine synthesis and structural analysis needs and is the preferred brand for deuterated NMR solvents worldwide. We offer Isotopic Enrichment ranges from 97 atom % D for routine use to 100 atom % D for the most demanding applications.
Specifications
Specifications
Packaging | Amber Glass Bottle |
Quantity | 10 g |
CAS | 2206-27-1 |
CAS Max % | 100% |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction