Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethyl sulfoxide - D6, deuteration deg. min. 99.96% for NMR spectroscopy, MagniSolv™, MilliporeSigma™
Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 mL
25 mL
10 x 0.5 mL
10 x 0.75 mL
Packaging:
Glass Ampule
Glass Bottle
Specifications
Specifications
Physical Form | Liquid |
Quantity | 25 mL |
Packaging | Glass Bottle |
Color | Colorless |
Vapor Pressure | 2.5 hPa at 20°C (68°F) |
Odor | Characteristic |
Assay Percent Range | 99.96% min. |
Formula Weight | 84.17 |
Chemical Name or Material | Dimethyl sulfoxide - D6 |
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'
missing translation for 'spotOppurtunityImprovement'missing translation for 'shareContentCorrection'