Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide - D6, deuteration degree min. 99.8% for NMR spectroscopy, MagniSolv™, MilliporeSigma™

Catalog No. M1034240010 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
50 mL
100 mL
10 x 0.5 mL
Packaging:
Glass Ampule
Glass Bottle
Septum Ampule
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1034240010 10 mL Septum Ampule
M1034240011 10 mL Glass Ampule
M1034240025 25 mL Glass Bottle
M1034240050 50 mL Glass Bottle
M1034240100 100 mL Glass Bottle
M1034240005 10 x 0.5 mL Glass Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. M1034240010 Supplier MilliporeSigma Supplier No. 1.03424.0010
Add to Cart
Add to Cart

For NMR Spectroscopy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-27-1
Molecular Formula C2H6OS
Molecular Weight (g/mol) 84.17
MDL Number MFCD00002090
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
PubChem CID 75151
IUPAC Name (²H₃)methanesulfinyl(²H₃)methane
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Quantity 10 mL
Color Colorless
Packaging Septum Ampule
Vapor Pressure 2.5 hPa at 20°C (68°F)
Odor Characteristic
Assay Percent Range 99.8% min.
Formula Weight 84.17
Chemical Name or Material Dimethyl sulfoxide - D6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.