Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide, HPLC Grade, 99.9+%

Catalog No. AA22914M1
Click to view available options
Quantity:
1 L
4 L
4 x 1 L

CAS: 67-68-5 | C2H6OS | 78.13 g/mol

Dimethyl sulfoxide is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Color Colorless
Appearance Colorless liquid
Density 1.101
Boiling Point 189°C
Quantity 4 x 1 L
Flash Point 87°C (189°F)
Ignition Point 270°C (518°F)
Odor Mild
Physical Form Liquid
Refractive Index 1.479
Assay Percent Range >99.9%
Beilstein 506008
Solubility Information Miscible with water,ethanol,ether,benzene,chloroform and acetone.
IUPAC Name methylsulfinylmethane
Formula Weight 78.13
Grade HPLC
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P370+P378q-P501c
EINECSNumber 200-664-3
RTECSNumber PV6210000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.