Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, Multi-Compendial, J.T.Baker™

Click to view available options
Quantity:
100mL
4L
19L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
Poly Pail

Chemical Identifiers

67-68-5
C2H6OS
78.129
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
CHEBI:28262
methylsulfinylmethane
CS(=O)C
Formula Weight 78.13
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.10kg
Flash Point 89.00°C
Quantity 100mL
Chemical Name or Material Dimethyl Sulfoxide
Grade Multi-Compendial