Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, Puriss p.a, ACS Reagent, ≥99.9% (GC), Honeywell Riedel-de Haën™

Catalog No. 60046839
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Poly Bottle

Puriss. p.a., ACS Reagent,≥99.9% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Color Colorless
Density 1.10g/mL
Quantity 2.5 L
Explosion Limit 42%
Ignition Point 573°C
Packaging Poly Bottle
Refractive Index 1.478 to 1.48
Linear Formula (CH3)2SO
Vapor Pressure 0.42mmHg (20°C)
Beilstein 506008
Water 0 to 0.1% (Coulometric)
IUPAC Name methanesulfinylmethane
Formula Weight 78.13g/mol
Percent Purity ≥99.9% (GC)
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
RTECSNumber 200-664-3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.