Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl terephthalate, 99%

Catalog No. AC180580025
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 120-61-6 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-61-6
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008440
InChI Key WOZVHXUHUFLZGK-UHFFFAOYSA-N
Synonym dimethyl terephthalate, dimethyl p-phthalate, 1,4-benzenedicarboxylic acid, dimethyl ester, dimethyl p-benzenedicarboxylate, di-me terephthalate, dimethyl 4-phthalate, terephthalic acid, dimethyl ester, dimethyl 1,4-benzenedicarboxylate, terephthalic acid dimethyl ester, methyl p-methoxycarbonyl benzoate
PubChem CID 8441
SMILES COC(=O)C1=CC=C(C=C1)C(=O)OC

Specifications

Acidity 0.03mg KOH/g max.
Melting Point 140°C to 143°C
Color White
Boiling Point 288°C
Flash Point 154°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 09, 843
Merck Index 15, 9305
Solubility Information Solubility in water: 0.036g/L (20°C). Other solubilities: soluble in ether and hot alcohol
Sublimation Point 285.0 °C
IUPAC Name dimethyl benzene-1,4-dicarboxylate
Formula Weight 194.19
Percent Purity 99%
Physical Form Flakes or Pellets
Chemical Name or Material Dimethyl terephthalate, 99%
Show More Show Less
EINECSNumber 204-411-8
RTECSNumber WZ1225000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.