Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl terephthalate, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
1kg
2.5kg
Packaging:
Plastic bottle
120-61-6
C10H10O4
194.19
MFCD00008440
WOZVHXUHUFLZGK-UHFFFAOYSA-N
dimethyl terephthalate,dimethyl p-phthalate,1,4-benzenedicarboxylic acid, dimethyl ester,dimethyl p-benzenedicarboxylate,di-me terephthalate,dimethyl 4-phthalate,terephthalic acid, dimethyl ester,dimethyl 1,4-benzenedicarboxylate,terephthalic acid dimethyl ester,methyl p-methoxycarbonyl benzoate
8441
dimethyl benzene-1,4-dicarboxylate
COC(=O)C1=CC=C(C=C1)C(=O)OC
120-61-6
C10H10O4
194.19
MFCD00008440
WOZVHXUHUFLZGK-UHFFFAOYSA-N
dimethyl terephthalate,dimethyl p-phthalate,1,4-benzenedicarboxylic acid, dimethyl ester,dimethyl p-benzenedicarboxylate,di-me terephthalate,dimethyl 4-phthalate,terephthalic acid, dimethyl ester,dimethyl 1,4-benzenedicarboxylate,terephthalic acid dimethyl ester,methyl p-methoxycarbonyl benzoate
8441
dimethyl benzene-1,4-dicarboxylate
COC(=O)C1=CC=C(C=C1)C(=O)OC
Boiling Point 288°C
Flash Point 154°C
Sublimation Point 285.0 °C
Infrared Spectrum Authentic
Quantity 25g
Packaging Plastic bottle
Acidity 0.03mg KOH/g max.
Melting Point 140°C to 143°C
Color White
Assay Percent Range 98.5% min. (GC)
Beilstein 09, 843
Merck Index 15, 9305
Solubility Information Solubility in water: 0.036g/L (20°C). Other solubilities: soluble in ether and hot alcohol
Formula Weight 194.19
Physical Form Flakes or Pellets
Percent Purity 99%
Chemical Name or Material Dimethyl terephthalate, 99%
Show More Show Less

EINECSNumber : 204-411-8

RTECSNumber : WZ1225000

TSCA : TSCA