Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Tetradecanedioate 96.0+%, TCI America™
SDP

Catalog No. p-7136930
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5024-21-5
Molecular Formula C16H30O4
Molecular Weight (g/mol) 286.41
MDL Number MFCD00673459
InChI Key ZDJFDFNNEAPGOP-UHFFFAOYSA-N
PubChem CID 78727
IUPAC Name 1,14-dimethyl tetradecanedioate
SMILES COC(=O)CCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 43°C
Color White
Boiling Point 196°C
Quantity 5 g
Synonym tetradecanedioic acid, dimethyl ester, 1,14-dodecanedicarboxylic acid dimethyl ester, tetradecanedioic acid dimethyl ester, tetradecanedioic acid, 1,14-dimethyl ester, acmc-1cupl, 1,14-dimethyl tetradecanedioate, tetradecanedioic acid,1,14-dimethyl ester
Formula Weight 286.41
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Tetradecanedioate
EINECSNumber (2)-2630
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.