Learn More
Dimethylamine, 40% w/w aq. soln., Thermo Scientific Chemicals
Description
Dimethylamine 40% aqueous solution is used as a precursor in the preparation of nitrosodimethylamine. It is also used in paint additives, lubricants, lubricant additives and coating additives. It plays a major role in ion exchange agents and agricultural chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.890 g/mL |
| Flash Point | 15°C (60°F) |
| Odor | Fish-like |
| Refractive Index | 1.37 |
| Quantity | 1 kg |
| UN Number | UN1160 |
| Beilstein | 605257 |
| Merck Index | 14,3228 |
| Solubility Information | Miscible with water and most organic solvents. |
| Formula Weight | 45.08 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302+H332-H314-H335 |
| Hazard Statement | GHS H Statement H225-H331-H314-H318-H302-H335 Highly flammable liquid and vapor. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIMETHYLAMINE SOLUTION |
| EINECSNumber | 204-697-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.