Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog No. AAA121330B
Click to view available options
Quantity:
250 g
1000 g
5000 g

Dimethylamine hydrochloride is used as an intermediate in the manufacture of pharmaceuticals like ranitidine and metformin, tramadol and amlodipine. It is used as a precursor of atrazine. It is associated with sodium acetate and used to carry out the Willgerodt- Kindler reaction to prepare amides. Its free base reacts with carbon disulfide to get dimethyldithiocarbamate which is used in rubber vulcanization. It is involved in the synthesis of dimethyl-(1-methyl-pyrrol-2-ylmethyl)-amine by reacting with 1-methyl pyrrole and formaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylamine hydrochloride is used as an intermediate in the manufacture of pharmaceuticals like ranitidine and metformin, tramadol and amlodipine. It is used as a precursor of atrazine. It is associated with sodium acetate and used to carry out the Willgerodt- Kindler reaction to prepare amides. Its free base reacts with carbon disulfide to get dimethyldithiocarbamate which is used in rubber vulcanization. It is involved in the synthesis of dimethyl-(1-methyl-pyrrol-2-ylmethyl)-amine by reacting with 1-methyl pyrrole and formaldehyde.

Solubility
Soluble in water, alcohol and chloroform

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-59-2
Molecular Formula C2H8ClN
Molecular Weight (g/mol) 81.543
MDL Number MFCD00012477
InChI Key IQDGSYLLQPDQDV-UHFFFAOYSA-N
Synonym dimethylamine hydrochloride, n-methylmethanamine hydrochloride, dimethylammonium chloride, methanamine, n-methyl-, hydrochloride, dimethylamine hcl, dimethylaminehydrochloride, unii-7m4cwb6aok, dimethylamine, hydrochloride, hydrochloric acid dimethylamine, n,n-dimethylamine hydrochloride
PubChem CID 10473
IUPAC Name N-methylmethanamine;hydrochloride
SMILES CNC.Cl

Specifications

Melting Point 168°C to 174°C
Quantity 1000 g
Beilstein 3589311
Sensitivity Hygroscopic
Merck Index 14,3228
Solubility Information Soluble in water,alcohol and chloroform
Formula Weight 81.55
Percent Purity ≥98%
Chemical Name or Material Dimethylamine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 208-046-5
RTECSNumber IQ0220000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.