Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylaminoethanol Bitartrate, Powder, 99%, Spectrum™ Chemical
SDP

Catalog No. 18610238 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
25 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-238 2.5 kg Poly Bottle
18-610-239 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-610-238 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D11752.5KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5988-51-2
Molecular Formula C8H17NO7
Molecular Weight (g/mol) 239.22
InChI Key UIEGYKVRCKDVKQ-UHFFFAOYNA-N
IUPAC Name 2,3-dihydroxybutanedioic acid; 2-(dimethylamino)ethan-1-ol
SMILES CN(C)CCO.OC(C(O)C(O)=O)C(O)=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 2.5 kg
Percent Purity 0.99
Grade Reagent
Physical Form Powder or Waxy Solid
Chemical Name or Material Dimethylaminoethanol Bitartrate

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.