Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2:1

Catalog No. ac292900250
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 4137-10-4 | C5H14N2O2 | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4137-10-4
Molecular Formula C5H14N2O2
Molecular Weight (g/mol) 134.18
MDL Number MFCD00029888
InChI Key JIYXHCMRGZVYMA-UHFFFAOYSA-N
Synonym DIMCARB
IUPAC Name dimethylamine; dimethylcarbamic acid
SMILES CNC.CN(C)C(O)=O

Specifications

Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 60°C to 61°C
Flash Point 13°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4520 to 1.456
Linear Formula ((CH3)2NH2)·(CH3)2NCO2)
Quantity 25 g
Specific Gravity 1.05
Solubility Information Solubility in water: may decompose. Other solubilities: forming carbon dioxide,dimethylamine
Viscosity 84.3 mPa.s (20°C)
Formula Weight 134.18
Physical Form Liquid
Chemical Name or Material Dimethylammonium-dimethylcarbamate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.