Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylcarbamyl chloride, 96%

Catalog No. aal0341514
Click to view available options
Quantity:
25 g
100 g

CAS: 79-44-7 | C3H6ClNO | 107.537 g/mol

Dimethylcarbamyl chloride is used to make dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylcarbamyl chloride is used to make dyes and pharmaceuticals.

Solubility
Reacts with water.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-44-7
Molecular Formula C3H6ClNO
Molecular Weight (g/mol) 107.537
MDL Number MFCD00000635
InChI Key YIIMEMSDCNDGTB-UHFFFAOYSA-N
Synonym dimethylcarbamoyl chloride, dimethylcarbamyl chloride, dimethylcarbamic chloride, dimethylcarbamoylchloride, carbamic chloride, dimethyl, dimethylcarbamic acid chloride, dimethyl carbamoyl chloride, n,n-dimethylcarbamyl chloride, carbamoyl chloride, dimethyl, dimethylcarbamidoyl chloride
PubChem CID 6598
ChEBI CHEBI:82280
IUPAC Name N,N-dimethylcarbamoyl chloride
SMILES CN(C)C(=O)Cl

Specifications

Melting Point -33°C
Boiling Point 167°C to 168°C
Refractive Index 1.453
Linear Formula (CH3)2NCOCl
UN Number UN2262
Beilstein 878197
Solubility Information Reacts with water.
Density 1.171
Quantity 25 g
Flash Point 68°C (154°F)
Odor Unpleasant
Formula Weight 107.54
Sensitivity Moisture sensitive
Percent Purity 96%
Chemical Name or Material Dimethylcarbamyl chloride
Show More Show Less
Hazard Category H227-H302-H315-H319-H331-H335-H350
Hazard Statement GHS H Statement
H331-H350-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause cancer.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P235-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 201-208-6
RTECSNumber FD4200000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.