Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 2,6-pyridinedicarboxylate, 99%

Catalog No. p-3770982
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle

CAS: 5453-67-8 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5453-67-8
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00134493
InChI Key SNQQJEJPJMXYTR-UHFFFAOYSA-N
Synonym dimethyl 2,6-pyridinedicarboxylate, 2,6-dimethyl pyridine-2,6-dicarboxylate, pyridine-2,6-dicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid, dimethyl ester, dimethyl dipicolinate, dimethyl pyridine-2,6-carboxylate, chembl72405, dimethylpyridine-2,6-dicarboxylate, dipicolinic acid dimethyl ester
PubChem CID 79549
IUPAC Name dimethyl pyridine-2,6-dicarboxylate
SMILES COC(=O)C1=NC(=CC=C1)C(=O)OC

Specifications

Melting Point 123°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 195.17
Percent Purity 99%
Chemical Name or Material Dimethyl 2, 6-pyridinedicarboxylate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 226-697-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.