Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl carbonate, 99%

Catalog No. p-3778373
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 616-38-6 | C3H6O3 | 90.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-38-6
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00008420
InChI Key IEJIGPNLZYLLBP-UHFFFAOYSA-N
Synonym methyl carbonate, carbonic acid, dimethyl ester, carbonic acid dimethyl ester, methyl carbonate meo 2co, unii-ke9j097spn, dimethylcarbonate, ch3ocooch3, ke9j097spn, dimethyl ester of carbonic acid, dsstox_cid_9192
PubChem CID 12021
ChEBI CHEBI:36596
IUPAC Name dimethyl carbonate
SMILES COC(=O)OC

Specifications

Melting Point 2.0°C to 4.0°C
Density 1.0690g/mL
Boiling Point 90.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3672 to 1.3692
Linear Formula (CH3O)2CO
Quantity 500 g
Beilstein 03, 4
Fieser 02,149; 05,234; 12,201
Specific Gravity 1.069
Merck Index 15, 3263
Solubility Information Solubility in water: 139g/L. Other solubilities: soluble in all proportions with organic alcohols,soluble in all proportions with esters and ketones,miscible with acids and alkalies
Viscosity 0.625 mPa.s (20°C)
Formula Weight 90.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl carbonate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-478-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.