Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Dimethyl L-tartrate, 98+%

Catalog No. AC172491000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 608-68-4 | C6H10O6 | 178.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-68-4
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00064437
InChI Key PVRATXCXJDHJJN-QWWZWVQMSA-N
Synonym +-dimethyl l-tartrate, dimethyl l-tartrate, 2r,3r-dimethyl 2,3-dihydroxysuccinate, dimethyl +-tartrate, l-+-tartaric acid dimethyl ester, dimethyl l-+-tartrate, 1,4-dimethyl 2r,3r-2,3-dihydroxybutanedioate, butanedioic acid, 2,3-dihydroxy-2r,3r-, 1,4-dimethyl ester, dimethyl 2r,3r-2,3-dihydroxybutanedioate, l-tartaric acid, dimethyl ester
PubChem CID 11851
IUPAC Name dimethyl (2R,3R)-2,3-dihydroxybutanedioate
SMILES COC(=O)C(C(C(=O)OC)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 48°C to 50°C
Color White
Boiling Point 157°C to 159°C (15.0 hPa)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98+% min. (GC)
Packaging Glass bottle
Linear Formula [-CH(OH)CO2CH3]2
Quantity 100 g
Beilstein 03, 510
Fieser 13,124
Specific Rotation Condition + 21.00 (20.00°C c=10,H2O)
Specific Rotation + 21
Solubility Information Solubility in water: soluble in water
Formula Weight 178.14
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material (+)-Dimethyl L-tartrate
Show More Show Less
EINECSNumber 210-166-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.