Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylformamide, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
HDPE Pressure Bottom Poly Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

68-12-2
C3H7NO
73.095
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 73.09
Packaging HDPE Pressure Bottom Poly Bottle
Specific Gravity 1L = 0.95kg
Flash Point 58.00°C
Quantity 500mL
Chemical Name or Material Dimethylformamide
Grade Baker Analyzed™ ACS Reagent