Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethylglyoxime, 1% Alcoholic, for Nickel, Certified, LabChem™
Click to view available options
Quantity:
125 mL
500 mL
Specifications
Specifications
Color | Yellow |
Boiling Point | 78°C |
Melting Point | -115°C |
CAS Max % | 1, 89.1, 4.95, 4.95 |
Linear Formula | CH3C(=NOH)C(=NOH)CH3 |
UN Number | UN1170 |
Density | 790kg/m3 |
Solubility Information | Soluble in water |
Formula Weight | 116.12 |
Grade | Certified |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement Highly flammable liquid and vapor. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Suspected of damaging fertility or the unborn child (oral). Causes damage to organs (central nervous system, optic nerve, liver, kidneys). |
Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Keep away from heat, hot surfaces, open flames, sparks. No smoking. Keep container tightly closed. Ground/bond container and receiving equipment. Use explosion-proof electrical, lighting, ventilating equipment. Use only non-sparking tools. Take precautionary measures against static discharge. Do not breathe mist, vapors, spray. Do not eat, drink or smoke when using this product. Use only outdoors or in a well-ventilated area. Wear eye protection, face protection, protective clothing, protective gloves. Wash exposed skin thoroughly after handling. If exposed or concerned: Get medical advice/attention. If swallowed: Rinse mouth. Call a poison center/doctor if you feel unwell. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If skin irritation occurs: Get medical advice/attention. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Call a poison center/doctor if you feel unwell. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish. Store locked up in a well-ventilated place. Dispose of contents/container to comply with local, state and federal regulations. |
Recommended Storage | Room Temperature |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction