Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride, 60% in methanol, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
Glass bottle
27668-52-6
C26H58ClNO3Si
496.29
MFCD00054224
WSFMFXQNYPNYGG-UHFFFAOYSA-M
quat-silsesquioxane,dmoap,dow corning 5700,caswell no. 892b,dimethyloctadecyl 3-trimethoxysilyl propyl ammonium chloride,unii-iq36o85wq4,n,n-dimethyl-n-3-trimethoxysilyl propyl octadecan-1-aminium chloride,epa pesticide chemical code 107401,3-trimethoxysilyl propyl dimethyl octadecyl ammonium chloride,3-trimethoxysilyl propyldimethyloctadecylammonium
62827
dimethyl-octadecyl-(3-trimethoxysilylpropyl)azanium;chloride
CCCCCCCCCCCCCCCCCC[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-]
27668-52-6
C26H58ClNO3Si
496.29
MFCD00054224
WSFMFXQNYPNYGG-UHFFFAOYSA-M
quat-silsesquioxane,dmoap,dow corning 5700,caswell no. 892b,dimethyloctadecyl 3-trimethoxysilyl propyl ammonium chloride,unii-iq36o85wq4,n,n-dimethyl-n-3-trimethoxysilyl propyl octadecan-1-aminium chloride,epa pesticide chemical code 107401,3-trimethoxysilyl propyl dimethyl octadecyl ammonium chloride,3-trimethoxysilyl propyldimethyloctadecylammonium
62827
dimethyl-octadecyl-(3-trimethoxysilylpropyl)azanium;chloride
CCCCCCCCCCCCCCCCCC[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-]
Phase transfer catalyst
Specific Gravity 0.89
Infrared Spectrum Authentic
Packaging Glass bottle
Density 0.8900g/mL
Quantity 100mL
Flash Point 15°C
Color Yellow
Linear Formula [(CH3O)3Si(CH2)3N(CH3)2(CH2)17CH3]Cl
Solubility Information Solubility in water: reacts
Formula Weight 496.29
Physical Form Solution
Percent Purity 60% in methanol
Chemical Name or Material Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly

GHS Signal Word: Danger

EINECSNumber : 248-595-8