Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylphenylsilane, 97%

Catalog No. p-7044099
Click to view available options
Quantity:
10 g
50 g

CAS: 766-77-8 | C8H11Si | 135.261 g/mol

Dimethylphenylsilane is a reagent for enol ether synthesis. It acts as a catalyst. Also used as a precursor in Optical Emission Spectroscopy of Plasma Deposition Processes. It can react with vinylbenzene to produce (b-Phenyl-ethyl)-dimethylphenyl-silan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylphenylsilane is a reagent for enol ether synthesis. It acts as a catalyst. Also used as a precursor in Optical Emission Spectroscopy of Plasma Deposition Processes. It can react with vinylbenzene to produce (b-Phenyl-ethyl)-dimethylphenyl-silan.

Solubility
Not miscible in water.

Notes
Moisture sensitive. Incompatible materials are Strong oxidizing agents, Strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-77-8
Molecular Formula C8H11Si
Molecular Weight (g/mol) 135.261
MDL Number MFCD00008257
InChI Key OIKHZBFJHONJJB-UHFFFAOYSA-N
Synonym dimethylphenylsilane, dimethyl phenyl silane, phenyldimethylsilane, silane, dimethylphenyl, benzene, dimethylsilyl, dimethylphenyl silane, c8h12si, dimethyl phenyl silyl, dimethyl-phenylsilicon
PubChem CID 6327656
IUPAC Name dimethyl(phenyl)silicon
SMILES C[Si](C)C1=CC=CC=C1

Specifications

Boiling Point 156°C to 157°C
Melting Point -124°C
Quantity 10 g
UN Number UN1993
Beilstein 2204906
Flash Point 35°C (95°F)
Solubility Information Not miscible in water.
Formula Weight 136.27
Percent Purity 97%
Refractive Index 1.497
Density 0.879
Chemical Name or Material Dimethylphenylsilane
Show More Show Less
Hazard Category H226-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 212-170-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.