Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylsulfamoyl Chloride, 99%

Catalog No. AC408591000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408591000 100 g Glass bottle
AC408590250 25 g Glass bottle
AC408595000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC408591000 Supplier Thermo Scientific Chemicals Supplier No. 408591000
Add to Cart

CAS: 13360-57-1 | C2H6ClNO2S | 143.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13360-57-1
Molecular Formula C2H6ClNO2S
Molecular Weight (g/mol) 143.59
MDL Number MFCD00007425
InChI Key JFCHSQDLLFJHOA-UHFFFAOYSA-N
Synonym dimethylsulfamoyl chloride, sulfamoyl chloride, dimethyl, dimethylaminosulfonyl chloride, dimethylsulfamoylchloride, dimethylsulphamoyl chloride, dimethylamidosulfonyl chloride, n,n-dimethylsulfamyl chloride, dimethylsulfamyl chloride, sulfamoyl chloride, n,n-dimethyl, dimethyl sulfamoyl chloride
PubChem CID 83372
IUPAC Name N,N-dimethylsulfamoyl chloride
SMILES CN(C)S(Cl)(=O)=O

Specifications

Melting Point -13°C
Density 1.3370g/mL
Boiling Point 114°C (75.0 mmHg)
Flash Point 94°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4510 to 1.453
Linear Formula (CH3)2NSO2Cl
Quantity 100 g
Beilstein 04, 84
Fieser 09,187
Specific Gravity 1.337
Solubility Information Solubility in water: decomposes in water
Formula Weight 143.59
Percent Purity 99%
Chemical Name or Material Dimethylsulfamoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
May cause cancer.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 236-412-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.