Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylthiocarbamoyl chloride, 99%

Catalog No. p-120724
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 16420-13-6 | C3H6ClNS | 123.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16420-13-6, 79-44-7
Molecular Formula C3H6ClNS
Molecular Weight (g/mol) 123.60
MDL Number MFCD00004919
InChI Key PHWISQNXPLXQRU-UHFFFAOYSA-N
Synonym dimethylthiocarbamoyl chloride, carbamothioic chloride, dimethyl, chloromethanethioyl dimethylamine, n,n-dimethylthiocarbamoyl chloride, n,n-dimethylcarbamothioic chloride, dimethylthiocarbamoylchloride, carbamoyl chloride, dimethylthio, carbamothioic chloride, n,n-dimethyl, dimethylthiocarbamyl chloride, dimethylthiocarbamic acid chloride
PubChem CID 27871
IUPAC Name N,N-dimethylcarbamothioyl chloride
SMILES CN(C)C(Cl)=S

Specifications

Melting Point 42°C to 43°C
Color Yellow
Boiling Point 63°C to 65°C (0.2 mbar)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCSCl
Quantity 25 g
Beilstein 04, 75
Fieser 02,173; 03,127; 04,202; 08,200
Solubility Information Solubility in water: reacts
Formula Weight 123.61
Percent Purity 99%
Physical Form Low Melting Crystalline Solid or Crystals
Chemical Name or Material Dimethylthiocarbamoyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause cancer.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 240-468-5
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.