Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethyltin dichloride, 95%
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 103.0°C to 110.0°C |
Color | White to Yellow |
Boiling Point | 188.0°C to 190.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (Argentometry) |
Packaging | Glass bottle |
Molecular Formula | C2H6Cl2Sn |
Linear Formula | (CH3)2Sn(Cl)2 |
Quantity | 25 g |
Solubility Information | Solubility in water: 20g/L (20°C). Other solubilities: 0.1 g/mL in methanol |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Toxic if swallowed. Toxic in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated |
Precautionary Statement | GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. |
EINECSNumber | 212-039-2 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction