Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIMT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117911
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5117911 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIPA5117911

Supplier: Invitrogen™ PA5117911

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DIMT1 Polyclonal Antibody for Western Blot, IHC (P), IP

DIMT1 specifically dimethylates two adjacent adenosines in the loop of a conserved hairpin near the 3'-end of 18S rRNA in the 40S particle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIMT1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene DIMT1
Gene Accession No. Q9UNQ2
Gene Alias 1500031M22Rik; 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase; 18S rRNA dimethylase; AV111347; DIM1; DIM1 dimethyladenosine transferase 1 homolog; DIM1 dimethyladenosine transferase 1 homolog (S. cerevisiae); DIM1 dimethyladenosine transferase 1-like; DIM1 dimethyladenosine transferase 1-like (S. cerevisiae); dimethyladenosine transferase; Dimt1; Dimt1l; HSA9761; HUSSY-05; HUSSY5; probable 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase; Probable 18S rRNA dimethylase; probable dimethyladenosine transferase; Probable S-adenosylmethionine-6-N',N'-adenosyl(rRNA) dimethyltransferase; RGD1311752; S-adenosylmethionine-6-N',N'-adenosyl(rRNA) dimethyltransferase
Gene Symbols DIMT1
Host Species Rabbit
Immunogen E. coli-derived Human DIMT1L fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27292
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.