Learn More
DIMT1L Mouse anti-Human, Clone: OTI2E6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810140

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
DIMT1 specifically dimethylates two adjacent adenosines in the loop of a conserved hairpin near the 3′-end of 18S rRNA in the 40S particle.Specifications
DIMT1L | |
Monoclonal | |
Unconjugated | |
DIMT1 | |
DIM1, DIMT1L, HSA9761, HUSSY5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
27292 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2E6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UNQ2 | |
DIMT1 | |
Human recombit protein fragment corresponding to amino acids 1-238 of human DIMT1L produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.