Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIMT1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 155631AP150UL

Description
DIMT1 specifically dimethylates two adjacent adenosines in the loop of a conserved hairpin near the 3′-end of 18S rRNA in the 40S particle.Specifications
DIMT1L | |
Polyclonal | |
Unconjugated | |
DIMT1 | |
18S rRNA dimethylase, DIM1, DIMT1, DIMT1L, HSA9761, HUSSY5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27292, 294718, 66254 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9D0D4, Q9UNQ2 | |
DIMT1 | |
DIMT1L Fusion Protein Ag7721 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction