Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dinaciclib
GREENER_CHOICESDP

Catalog No. 733650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-365-0 50 mg
73-361-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 73-365-0 Supplier Tocris Bioscience™ Supplier No. 7336/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective inhibitor of CDK2, CDK5, CDK1 and CDK9

Dinaciclib is a potent and selective inhibitor of CDK2, CDK5, CDK1 and CDK9 activity in vitro (IC50 values are 1, 1, 3 and 4 nM, respectively). The compound inhibits DNA replication by blocking thymidine DNA incorporation in A2780 cells (IC50 = 4 nM). Dinaciclib induces regression of established solid tumors in mouse models and shows antiviral activity against SARS-CoV-2 in vitro (Vero E6, IC50 = 0.127 μM; A549-ACE2, IC50 = 0.032 μM).

Specifications

Chemical Name or Material (2 S)-1-[3-Ethyl-7-[[(1-oxido-3-pyridinyl)methyl]amino]pyrazolo[1,5-a]pyrimidin-5-yl]-2-piperidineethanol
CAS 779353-01-4
Quantity 50 mg
Target Cyclin-dependent Kinase Inhibitors
Molecular Formula C21H28N6O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.