Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dinonyl phthalate, mixture of isomers, 96%

Catalog No. AAA1287118
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1287118 50 g
AAA1287109 10 g
AAA1287130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1287118 Supplier Thermo Scientific Chemicals Supplier No. A1287118
Add to Cart

CAS: 84-76-4 | C26H42O4 | 418.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-76-4
Molecular Formula C26H42O4
Molecular Weight (g/mol) 418.62
MDL Number MFCD00036237
InChI Key DROMNWUQASBTFM-UHFFFAOYSA-N
Synonym dinonyl phthalate, dinonylphthalate, nonyl phthalate, di-n-nonyl phthalate, bisoflex dnp, unimoll dn, bisoflex 91, phthalic acid, dinonyl ester, 1,2-benzenedicarboxylic acid, dinonyl ester, bisolflex 91
PubChem CID 6787
IUPAC Name dinonyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC

Specifications

Density 0.97
Flash Point 216°C (421°F)
Quantity 50 g
Beilstein 1916263
Formula Weight 418.62
Percent Purity 98%
Chemical Name or Material Dinonyl phthalate, mixture of isomers
EINECSNumber 201-560-0
RTECSNumber TI1800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.