Learn More
Invitrogen™ DIO1 Monoclonal Antibody (1E4)

Description
Peptide Sequence: DRVKRNILAM GEKTGMTRNP HFSHDNWIPT FFSTQYFWFV LKVRWQRLED TTELGGLAPN CPVVRLSGQR CNIWEFMQGN RPLVLNFGSC T.
Specifications
Specifications
| Antigen | DIO1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1E4 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Dio1 |
| Gene Accession No. | P49895 |
| Gene Alias | 5DI; D1; deiodinase, iodothyronine type I; deiodinase, iodothyronine, type I; Dio1; DIOI; hDido1; iodothyronine deiodinase type 1; ITDI1; thyroxine 5'-deiodinase; thyroxine deiodinase; thyroxine deiodinase type I (selenoprotein); thyroxine deiodinase, type I; TXDI1; Type 1 DI; type I iodothyronine deiodinase; Type-I 5'-deiodinase |
| Gene Symbols | Dio1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.