Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501727457 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7457 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7457 Supplier Proteintech Group Inc Supplier No. 117901AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a thiol-requiring propylthiouracil-sensitive oxidoreductase. It activates thyroid hormone by converting the prohormone thyroxine (T4) by outer ring deiodination (ORD) to bioactive 3,3′,5-triiodothyronine (T3). It also degrades both hormones by inner ring deiodination (IRD). Alternative splicing results in multiple transcript variants encoding different isoforms. Some, but not all, isoforms contain a selenocysteine (Sec) residue encoded by the UGA codon, which normally signals translation termination. The 3′ UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Additional transcript variants have been described but are not supported by experimental evidence.

Specifications

Antigen DIO1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene DIO1
Gene Accession No. P24389, P49895, Q61153
Gene Alias 5DI, DIO1, DIOI, ITDI1, TXDI1, Type 1 DI, Type I 5 deiodinase
Gene Symbols DIO1
Host Species Rabbit
Immunogen DIO1 Fusion Protein Ag2410
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13370, 1733, 25430
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.