Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIO2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549631
Description
Predicted to react with pig based on sequence homology.
The protein encoded by this gene belongs to the iodothyronine deiodinase family. It activates thyroid hormone by converting the prohormone thyroxine by outer ring deiodination to bioactive 3,3',5-triiodothyronine residues encoded by the UGA codon, which normally signals translation termination. The 3' UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence, which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
DIO2 | |
Polyclonal | |
Unconjugated | |
DIO2 | |
5DII; AI324267; D2; deiodinase, iodothyronine, type 2; deiodinase, iodothyronine, type II; deiodinase-2; deiodonase-2; Dio2; DIOII; iodothyronine type II deiodinase; Itdi2; SelY; thyroxine deiodinase; thyroxine deiodinase, type II; thyroxine type II; TXDI2; Type 2 DI; type 2 iodothyronine deiodinase; type II iodothyronine deiodinase; type-II 5' deiodinase; type-II 5'deiodinase; type-II 5'-deiodinase | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
13371, 1734 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q92813, Q9Z1Y9 | |
DIO2 | |
KLH conjugated synthetic peptide between 165-191 amino acids of human DIO2. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction