Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIO3 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP105767AF647
Description
DIO3 Polyclonal specifically detects DIO3 in Human, Mouse, Rat, Hamster, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
DIO3 | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
5DIII, D3, deiodinase, iodothyronine, type III, DIOIII, EC 1.97.1, EC 1.97.1.11, ITDI3, placental type, thyroxine deiodinase type III (selenoprotein), Type 3 DI, type 3 iodothyronine selenodeiodinase, type III iodothyronine deiodinase, type-III 5' deiodinase, Type-III 5'-deiodinase | |
Rabbit | |
31 kDa | |
0.1 mL | |
Cancer, Growth and Development, Lipid and Metabolism, Neuroscience, Signal Transduction | |
1735 | |
Human, Mouse, Rat, Hamster, Rabbit | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DIO3 | |
Synthetic peptide made to an internal portion of rat DIO3 (within residues 250-300). [Swiss-Prot# P49897] | |
Affinity Purified | |
RUO | |
Primary | |
Human, rat, hamster and mouse. Rabbit reactivity via customer review. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction