Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIO3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA526537

Catalog No. PIPA526537


Only null left
Add to Cart

Description

Description

DIO3 Polyclonal Antibody for Western Blot, IHC (P)

DIO3 belongs to the iodothyronine deiodinase family. It catalyzes the inactivation of thyroid hormone by inner ring deiodination of the prohormone thyroxine (T4) and the bioactive hormone 3,3',5-triiodothyronine (T3) to inactive metabolites, 3,3',5'-triiodothyronine (RT3) and 3,3'-diiodothyronine (T2), respectively. This enzyme is highly expressed in the pregnant uterus, placenta, fetal and neonatal tissues, suggesting that it plays an essential role in the regulation of thyroid hormone inactivation during embryological development. This protein contains a selenocysteine (Sec) residue, which is essential for efficient enzyme activity. The selenocysteine is encoded by the UGA codon, which normally signals translation termination. The 3' UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DIO3
Polyclonal
Unconjugated
DIO3
5DIII; D3; deiodinase, iodothyronine type III; deiodinase, iodothyronine, type 3; deiodinase, iodothyronine, type III; Dio3; DIOIII; I79_015795; iodothyronine deiodinase 3; iodothyronine deiodinase, placental type; Itdi3; LOW QUALITY PROTEIN: thyroxine 5-deiodinase; thyroxine 5-deiodinase; thyroxine deiodinase; thyroxine deiodinase type III (selenoprotein); TXDI3; Type 3 DI; type 3 iodothyronine selenodeiodinase; type III iodothyronine deiodinase; type-III 5' deiodinase; type-III 5'-deiodinase
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
1735, 29475
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P49897, P55073
DIO3
KLH conjugated synthetic peptide between 250-278 amino acids from the C-terminal region of human DIO3.
400 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.