Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dio3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985740
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5740 Supplier Bioss Supplier No. BS3902R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DIO3 belongs to the iodothyronine deiodinase family. It catalyzes the inactivation of thyroid hormone by inner ring deiodination of the prohormone thyroxine (T4) and the bioactive hormone 3,3′,5-triiodothyronine (T3) to inactive metabolites, 3,3′,5′-triiodothyronine (RT3) and 3,3′-diiodothyronine (T2), respectively. This enzyme is highly expressed in the pregnant uterus, placenta, fetal and neonatal tissues, suggesting that it plays an essential role in the regulation of thyroid hormone inactivation during embryological development. This protein contains a selenocysteine (Sec) residue, which is essential for efficient enzyme activity. The selenocysteine is encoded by the UGA codon, which normally signals translation termination. The 3′ UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal.

Specifications

Antigen Dio3
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DIO3
Gene Accession No. P55073
Gene Alias 5DIII; D3; deiodinase, iodothyronine type III; deiodinase, iodothyronine, type 3; deiodinase, iodothyronine, type III; Dio3; DIOIII; I79_015795; iodothyronine deiodinase, placental type; ITDI3; Thyroxine 5-deiodinase; thyroxine deiodinase type III (selenoprotein); Txdi3; Type 3 DI; type 3 iodothyronine selenodeiodinase; Type III iodothyronine deiodinase; type-III 5′ deiodinase; type-III 5′-deiodinase
Gene Symbols DIO3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Dio3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1735
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.