Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dioctyl Maleate (Diethylhexyl Maleate), 95+%, Neat Compound, Ultra Scientific

Catalog No. usdmp036
Encompass
Click to view available options
Quantity:
100 mg

Chemical Identifiers

CAS 142-16-5
Molecular Formula C20H36O4
Molecular Weight (g/mol) 340.504
InChI Key ROPXFXOUUANXRR-YPKPFQOOSA-N
PubChem CID 5365125
IUPAC Name bis(2-ethylhexyl) (Z)-but-2-enedioate
SMILES CCCCC(CC)COC(=O)C=CC(=O)OCC(CC)CCCC

Specifications

CAS Max % 100.0
Assay Percent Range >95%
Physical Form Liquid
Quantity 100 mg
Chemical Name or Material Dioctyl Maleate (Diethylhexyl Maleate)
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.