Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dioctyl Phthalate, Reagents

Catalog No. 01337276
Change view
Click to view available options
:
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-337-276 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-276 Supplier Reagents Holdings Llc Supplier No. C21971004C
Only null left
Add to Cart
Add to Cart

Dioctyl Phthalate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 117-81-7
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.56
InChI Key BJQHLKABXJIVAM-UHFFFAOYNA-N
Synonym Bis(2-ethylhexyl) Phthalate
IUPAC Name 1,2-bis(2-ethylhexyl) benzene-1,2-dicarboxylate
SMILES CCCCC(CC)COC(=O)C1=CC=CC=C1C(=O)OCC(CC)CCCC

Specifications

Boiling Range 384°C
Color Colorless
Boiling Point 384°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 0.9861
Grade Reagent
Physical Form Liquid
Chemical Name or Material Dioctyl Phthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.