Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dioctyl sulfosuccinate sodium salt, ∽99%, MP Biomedicals™

Catalog No. ICN10157590 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10157590 500 g
ICN10157550 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10157590 Supplier MP Biomedicals, Inc Supplier No. 0210157590
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 577-11-7
Molecular Formula C20H37NaO7S
Molecular Weight (g/mol) 444.56
MDL Number MFCD00012455
InChI Key APSBXTVYXVQYAB-UHFFFAOYNA-M
Synonym docusate sodium, dioctyl sodium sulfosuccinate, aerosol ot, constonate, diox, manoxol ot, diomedicone, clestol, complemix, defilin
PubChem CID 23673837
IUPAC Name sodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate
SMILES [Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

Specifications

Color White
Assay Percent Range ∽99%
Quantity 500 g
Formula Weight 444.57
Grade Not Available
Physical Form Solid
Chemical Name or Material Dioctyl sulfosuccinate sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.