Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Diorite Specimen
SCI_ED

Catalog No. S26670 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S26670 1 Ea. 7 to 8 cm
S24750 1 Ea. 2 to 3 cm
S26632 10/Pk. 2 to 3 cm
S24755 12/Pk. 2 to 3 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S26670 Supplier Eisco™ Supplier No. ESNG0042HAND
Add to Cart
Add to Cart

Diorite Specimen straddles the line between mafic and felsic rock chemistry.

  • Presence of both mafic and felsic minerals make diorite example of intermediate igneous chemistry
  • For use in lessons in bulk chemistry
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Large crystal size accommodates mineral identification with hand lens (not included)
  • The minerals within diorite can be readily identified due to their large size

Specifications

Product Type Igneous Rock
Quantity 1 Ea.
Specimen Size 7 to 8 cm
Weight (Metric) 0.5 kg
Specimen Type Diorite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.