Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diosmin 85.0+%, TCI America™
SDP

Catalog No. D390825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D390825G 25 g
D39085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D390825G Supplier TCI America Supplier No. D390825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 520-27-4
Molecular Formula C28H32O15
Molecular Weight (g/mol) 608.549
MDL Number MFCD00009772
InChI Key GZSOSUNBTXMUFQ-YFAPSIMESA-N
Synonym diosmin, barosmin, diosmine, venosmine, diosmil, flebosten, daflon, diosimin, diosminum, tovene
PubChem CID 5281613
ChEBI CHEBI:4631
IUPAC Name 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
SMILES CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=CC(=C4C(=C3)OC(=CC4=O)C5=CC(=C(C=C5)OC)O)O)O)O)O)O)O)O

Specifications

Melting Point 280°C
Quantity 25 g
Formula Weight 608.55
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Diosmin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.