Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Diotame Tablets

Catalog No. p-7078229
Click to view available options
Quantity:
2/Pk., 250/Box
2/Pk., 50/Box

Used as antacid and diarrhea relief.

  • Sugar-free
  • Soothes irritation with protective coating action
  • Each tablet contains Bismuth Subsalicylate 262mg

Specifications

Active Ingredients Bismuth Subsalicylate (262 mg)
For Use With (Application) Antacid and diarrhea-relief
Product Type Diotame Tablets
Color Pink
Format Sugar-free Tablet
Quantity 2/Pk., 50/Box
Recommended Age 12 years and older
Storage Requirements 59°F to 86°F (15°C to 30°C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.