Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Dip Stain Kits (Wright and Wright Giemsa)

Catalog No. p-4907645
Click to view available options
Quantity:
4 x 3800 mL (1 gal.)
4 x 950 mL
Type:
Wright Dip Stain Kit
Wright Giemsa Dip Stain Kit

For Hemotology staining using Dip Stain Kit (Wright & Wright Giemsa). Kit includes Solution I - Fixative, Solution II - Wright or Wright Giemsa Dip Stain, Solution III - Matching Buffer, and Solution IV - Alcohol Wash.

  • Solutions made from certified dyes
  • Solutions should be replenished during staining period
  • During cold weather, allow solutions to return to room temperature and shake well before use
  • Dip Stain Kit includes: Wright or Wright Giemsa Dip Stain, methanol, buffers, stabilizers
  • Dip Stain Kit Replacement Solutions include: 1 gallon of solution
  • For in-vitro diagnostic use only

Specifications

Quantity 4 x 3800 mL (1 gal.)
Storage Requirements Room Temperature
Type Wright Giemsa Dip Stain Kit
For Use With (Application) For in-vitro diagnostic use only
Packaging Quantity 1 Kit
Packaging Complete Kit (1 each Solutions I - IV)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.