Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIP13B Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145203
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145203 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145203 Supplier Invitrogen™ Supplier No. PA5145203
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELAKQLADTM VLPIIQFREK DLTEVSTLKD LFGLASNEHD LSMAKYSRLP KKKENEKVKT EVGKEVAAAR RKQHLSSLQY YCALNALQYR KQMA Antigen sequence identity: Mouse 93%; Rat 93%.

Required for the regulation of cell proliferation in response to extracellular signals mediated by an early endosomal compartment. Links Rab5 to nuclear signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIP13B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APPL2
Gene Accession No. Q8NEU8
Gene Alias adapter protein containing PH domain, PTB domain and leucine zipper motif 2; adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2; adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2; Appl2; DCC-interacting protein 13-beta; DIP13 beta; DIP13B; dip13-beta; Dip3 beta; Dip3b; RGD1563028
Gene Symbols APPL2
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 102-195. Recombinant protein control fragment (Product #RP-109744).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55198
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.