Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIP2B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522089

Catalog No. PIPA522089


Only null left
Add to Cart

Description

Description

Recommended positive controls: HeLa, Molt-4, mouse brain. Predicted reactivity: Mouse (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the disco-interacting protein homolog 2 protein family. The encoded protein contains a binding site for the transcriptional regulator DNA methyltransferase 1 associated protein 1 as well as AMP-binding sites. The presence of these sites suggests that the encoded protein may participate in DNA methylation. This gene is located near a folate-sensitive fragile site, and CGG-repeat expansion in the promoter of this gene which affects transcription has been detected in individuals containing this fragile site on chromosome 12.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DIP2B
Polyclonal
Unconjugated
DIP2B
4932422C22; AI317237; AI854602; DIP2 disco-interacting protein 2 homolog B; DIP2 disco-interacting protein 2 homolog B (Drosophila); DIP2 homolog B; Dip2b; disco interacting protein 2 homolog B; disco-interacting protein 2 homolog B; KIAA1463; mKIAA1463
Rabbit
Antigen affinity chromatography
RUO
239667, 57609
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q3UH60, Q9P265
DIP2B
Recombinant protein encompassing a sequence within the C-terminus region of human DIP2B. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.