Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIP2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561159
Description
Immunogen sequence: EKKRSKLLSP YSPQTQETDS AVQKELRNQT PAPSAAQTSA PSKYHRTRSG GARDERY Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
This gene encodes a member of the disco-interacting protein homolog 2 protein family. The encoded protein contains a binding site for the transcriptional regulator DNA methyltransferase 1 associated protein 1 as well as AMP-binding sites. The presence of these sites suggests that the encoded protein may participate in DNA methylation. This gene is located near a folate-sensitive fragile site, and CGG-repeat expansion in the promoter of this gene which affects transcription has been detected in individuals containing this fragile site on chromosome 12.
Specifications
DIP2B | |
Polyclonal | |
Unconjugated | |
DIP2B | |
4932422C22; AI317237; AI854602; DIP2 disco-interacting protein 2 homolog B; DIP2 disco-interacting protein 2 homolog B (Drosophila); DIP2 homolog B; Dip2b; disco interacting protein 2 homolog B; disco-interacting protein 2 homolog B; KIAA1463; mKIAA1463 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57609 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P265 | |
DIP2B | |
Recombinant protein corresponding to Human DIP2B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction