Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIP2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563241
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563241 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563241 Supplier Invitrogen™ Supplier No. PA563241
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IGEICVSSRT GGMMYFGLAG VTKNTFEVIP VNSAGSPVGD VPFIRSGLL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

This gene encodes a member of the disco-interacting protein homolog 2 protein family. The encoded protein contains a binding site for the transcriptional regulator DNA methyltransferase 1 associated protein 1 as well as AMP-binding sites. The presence of these sites suggests that the encoded protein may participate in DNA methylation. This gene is located near a folate-sensitive fragile site, and CGG-repeat expansion in the promoter of this gene which affects transcription has been detected in individuals containing this fragile site on chromosome 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIP2B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DIP2B
Gene Accession No. Q9P265
Gene Alias 4932422C22; AI317237; AI854602; DIP2 disco-interacting protein 2 homolog B; DIP2 disco-interacting protein 2 homolog B (Drosophila); DIP2 homolog B; Dip2b; disco interacting protein 2 homolog B; disco-interacting protein 2 homolog B; KIAA1463; mKIAA1463
Gene Symbols DIP2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DIP2B. Recombinant protein control fragment (Product #RP-103243).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57609
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.