Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIP2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566248 Supplier Invitrogen™ Supplier No. PA566248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKRSKLIGAYL PQPPRVDQAL PQERRAPVTP SSASRYHRRR SSGSRDERY Highest antigen sequence identity to the following orthologs - mouse 100%, rat 68%.

This gene encodes a member of the disco-interacting protein homolog 2 family. The protein shares strong similarity with a Drosophila protein which interacts with the transcription factor disco and is expressed in the nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIP2C
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DIP2C
Gene Accession No. Q9Y2E4
Gene Alias 2900024P20Rik; 9630044M06; DIP2 disco-interacting protein 2 homolog C; DIP2 disco-interacting protein 2 homolog C (Drosophila); DIP2 homolog C; DIP2C; disco interacting protein 2 homolog C; disco-interacting protein 2 homolog C; KIAA0934; mKIAA0934; RGD1560155
Gene Symbols DIP2C
Host Species Rabbit
Immunogen Recombinant Human DIP2C. Recombinant protein control fragment (Product #RP-105126).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22982
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.